Order Entry
Puerto Rico
ContactUsLinkComponent
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
Catalog # TS35884-2500
CAS Number:  
undefined
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
Catalog # TS35884-2500
Supplier Number:  35884-2500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 mg
  • Cat. No.:
    TS35884-2500

Specifications

Specification Test Results

  • Appearance:
    Light yellow Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥95 %

Specification Test Results

About this item

 :
 :187.43 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:75705-21-4
Formula:C₇H₁₁BClNO₂
MDL Number:MFCD01632199