ContactUsLinkComponent
Eisco Aluminum Cylindrical Water Bath
Eisco Aluminum Cylindrical Water Bath
Catalog # 470345-130
Supplier:  Wards
undefined
Eisco Aluminum Cylindrical Water Bath
Catalog # 470345-130
Supplier:  Wards
Supplier Number:  CH0106A
Specifications
  • Cat. No.:
    470345-130
Specifications
About this item
Provides maximum volume with smallest footprint.
  • Aluminum construction for higher heat dissipation
  • Lid is comprised of a set of concentric rings
  • Features two side handles and a top handle for safe and easy handling
Cylindrical water bath made of thick aluminum sheet, which is ideal for the best heat dissipation. The lid is comprised of a set of concentric rings. Capacity: 100 ml ; size: approximately 4" Ø (100 mm) with one pack size.