Eisco Teclu Burner, Natural Gas
Catalog # 470344-720
Supplier: Wards
Specifications
- Cat. No.:470344-720
Specifications
About this item
Produces very hot flame suitable for sterilization, glasswork, and smoldering.
- Suitable for use with Natural Gas (methane)
- Tapered riffled tube connector arm accepts 6 to 8 mm tubing
- Aluminum mixing tube is mounted on a circular, die casted zinc alloy base
- Needle valve allows for fine flame adjustment
- Unique spring operated gas safety lock is incorporated for air mix control
Premium quality Teclu burner that allows you to control both the air mix adjustment and gas flow adjustment. Burner produces a very hot flame and soldering. Burner measures 6.1" tall, and the base measures 3" in Ø.