
4-Nitro-1H-pyrazole 97%
Catalog # TS43016-0050
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:5 g
- Cat. No.:TS43016-0050
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:160 to 165 °C
- HPLC:≥96.0 %
- UV transmittance:≥99 % (c=0.01M water, 254 nm)
Specification Test Results
About this item
:
:113.0800 g/mol
Melting Pt:161…162 °C
:GHS07
Storage Temperature:Ambient
CAS Number:2075-46-9
Formula:C₃H₃N₃O₂
MDL Number:MFCD00159626