Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
1-Boc-4-piperidone 99%

1-Boc-4-piperidone 99%

Irritant
1-Boc-4-piperidone 99%
Certificates
1-Boc-4-piperidone 99%

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone
  • CAS Number:79099-07-3
  • Formula:C₁₀H₁₇NO₃
  • MDL Number:MFCD00151800
  • Molecular Weight:199.25 g/mol
  • Melting Pt:74…75 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    73 to 77 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    5 g
    Discontinued
    Supplier Number: -
    Specifications:
  • Glass bottle
    25 g
    Discontinued
    Supplier Number: 33027-0250
    Specifications:
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $503.00
    $503.00
    Each (100g)
       $503.00

    Supplier Number: 33027-1000
    Specifications:

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.