
Pyridine-2,5-dicarboxylic acid 98%
Catalog # TS13187-0050
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle for solids
- Size:5 g
- Cat. No.:TS13187-0050
Specifications
Specification Test Results
- Appearance:Yellow to green Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥97.5 %
- Sulfate (SO4):≤0.005 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.1 % (in water)
Specification Test Results
About this item
:
:167.12 g/mol
Merck Index:13,05177
Boiling Pt:357 °C (1013 hPa)
Melting Pt:256 °C (dec.)
Storage Temperature:Ambient
EINECS:202-834-2
:GHS07
CAS Number:100-26-5
Formula:C₇H₅NO₄
MDL Number:MFCD00006297