Formaldehyde 37%
Supplier: StatLab
Jump to:
About this item
37% Formaldehyde solution.
- Contains methanol as a stabilizer
Part of the StatLab line of prefilled bulk containers
Manufactured in the USA
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- UN:2209
- ADR:8,III
1 Options Available Below
Product Details & Documents
37% Formaldehyde solution.
- Contains methanol as a stabilizer
Part of the StatLab line of prefilled bulk containers
Manufactured in the USA
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- UN:2209
- ADR:8,III



