Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Formalin 10% Modified Carson/Millonig

Cube, 5 gallon
Catalog # 76548-034
Corrosive
Irritant
Health
Supplier:   StatLab
 :  28610-5
Formalin 10% Modified Carson/Millonig
Catalog # 76548-034
Supplier:  StatLab
CAS Number:  50-00-0
Formalin 10% Modified Carson/Millonig
Formalin 10% Modified Carson/Millonig

Cube, 5 gallon

CAS Number:  50-00-0
Price
$66.88
Log in for availability
Each (5US Gal)
Product Details & Documents

Formula preserves tissue specimens

  • The Carson Millonig modified buffering formulation preserves tissue specimens for both light and electron microscopy
  • Can be used in place of traditional 10% NBF

Part of the StatLab line of prefilled bulk containers

Manufactured in the USA

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
Specifications

Specifications

  • Pack type:
    Cube
  • Size:
    5 gallon

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.