Formalin 10% Modified Carson/Millonig
Supplier: StatLab
Jump to:
About this item
A modified buffering formulation to preserve tissue.
- Carson Millonig modified buffering formulation preserves tissue specimens
- For both light and electron microscopy
- Can be used in place of traditional 10% NBF
Part of the StatLab line of StatClick 10% NBF Prefills.
Manufactured in the USA
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
4 Options Available Below
Product Details & Documents
A modified buffering formulation to preserve tissue.
- Carson Millonig modified buffering formulation preserves tissue specimens
- For both light and electron microscopy
- Can be used in place of traditional 10% NBF
Part of the StatLab line of StatClick 10% NBF Prefills.
Manufactured in the USA
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: CM1507
- Supplier Number: CM1115
- Supplier Number: CM1230
- Supplier Number: CM1460



