
4-(Aminomethyl)phenol hydrate 97%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:696-60-6
:
Formula:C₇H₉NO·xH₂O
MDL Number:MFCD19690971
:123.16 g/mol (anhy)
Melting Pt:100…104 °C
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Off-white to light yellow Powder
- Infrared spectrum:Conforms
- Water:≤13.0 %
- Elemental Analysis:57.18 to 61.94 % (C) based on monohydrate
- UV:at 220 nm A: ≤0.06
- Titration after Ion exchange:≥99.0 % (on dry substance)
- Membrane filtration test:(on 0.5 M soln.) Passes test
- Suitability:(HPLC gradient) Passes test
Specification Test Results
Frequently Bought Together