
4-(Trifluoromethyl)thiophenol 97%
Catalog # TS35529-0010
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle for solids
- Size:1 g
- Cat. No.:TS35529-0010
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4920 to 1.4950 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:178.18 g/mol
Boiling Pt:68…72 °C (15 mmHg)
:GHS07
Storage Temperature:Refrigerator
CAS Number:825-83-2
Formula:C₇H₅F₃S
MDL Number:MFCD00042320