

2-Naphthol 99+%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:135-19-3
:
Formula:C₁₀H₈O
MDL Number:MFCD00004067
Merck Index:12,06471
Boiling Pt:285…286 °C (760 mmHg)
Melting Pt:120…124 °C
Flash Pt:153 °C
Storage Temperature:Ambient
EINECS:205-182-7
ADR:9,III
:144.17 g/mol
UN:3077
:GHS07
:GHS09
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White to beige to pink to light brown Powder or flakes
- Infrared spectrum:Conforms
- Melting point:120 to 124 °C
- GC:≥99.0 %