Order Entry
Puerto Rico
ContactUsLinkComponent
Organic Reference Standard, Caffeine (1 mg/ml) in Methanol
Organic Reference Standard, Caffeine (1 mg/ml) in Methanol
Catalog # TS39354-0010
CAS Number:  
Organic Reference Standard, Caffeine (1 mg/ml) in Methanol
Catalog # TS39354-0010
Supplier Number:  39354-0010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • CAS No.:
    58-08-2
  • Description:
    Caffeine (< 100 µg/ml) in methanol
  • Formula:
    C₈H₁₀N₄O₂
  • Description:
    Caffeine
  • Cat. No.:
    TS39354-0010

Specifications

About this item