Order Entry
Puerto Rico
ContactUsLinkComponent
 
Health
Formamide, dry, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Catalog # BJ34724-1L
CAS Number:  
Health
Formamide, dry, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Catalog # BJ34724-1L
Supplier Number:  34724-6X1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    6 x 1 L
  • Cat. No.:
    BJ34724-1L
  • Supplier no.:
    34724-6X1L

Specifications

Specification Test Results

  • Suitability (for KF-titration):
    test passed
  • Water (Karl Fischer):
    Max. 0.02 %
  • Number Of Samples:
    Min. 2
  • Vapor Density:
    1.55 (vs air)
  • Refractive Index:
    n20/D 1.447 (lit.)
  • Vapor Pressure:
    30 mmHg ( 129 °C)
  • Autoignition Temperature:
    932 °F

Specification Test Results

About this item

 :
Merck Index:13,04261
Boiling Pt:210 °C (1.013 hPa)
Melting Pt:1…3 °C
Density:1.133 g/cm³ (20 °C)
Flash Pt:175 °C
EINECS:200-842-0
ADR:1,
 :45.04 g/mol
 :GHS08
Storage Temperature:Room Temperature
CAS Number:75-12-7
Formula:HCONH₂
MDL Number:MFCD00007941