SARS-CoV-2 Nucleocapsid protein (NP) High-sensitivity Quantitative ELISA, BioVendor
Catalog # 103853-660
Supplier: BioVendor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Direct
- Conjugate ELISA:HRP
- Format:Pre-coated
- Host:Human
- Primary antibody reactivity:Human
- Target protein:anti-SARS-CoV-2
nucleocapsid protein - Description:SARS-CoV-2 Nucleocapsid protein (NP) High-sensitivity Quantitative ELISA
- Size:1 kit
- Sample Type:Serum, Plasma
- Detection Method:Colorimetric
- Shelf Life:See expiration date
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 µl
- Regulatory Status:RUO
- Cat. No.:103853-660
Specifications
About this item
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of anti-SARS-CoV-2
nucleocapsid protein.
Caution: This is intended for research use only.