Order Entry
Puerto Rico
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Diameter ~2 mm, Fluka™
Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Diameter ~2 mm, Fluka™
Catalog # BJ34241-250G
CAS Number:  
undefined
Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Diameter ~2 mm, Fluka™
Catalog # BJ34241-250G
Supplier Number:  34241-250G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 x 250 g
  • Cat. No.:
    BJ34241-250G
  • Supplier no.:
    34241-250G

Specifications

Specification Test Results

  • Suitability (for KF-titration):
    test passed

Specification Test Results

About this item

 :
Density:2 g/cm³
Storage Temperature:Ambient
CAS Number:308080-99-1
 :1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613