Order Entry
Puerto Rico
ContactUsLinkComponent
Orthophosphoric acid ≥85%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Corrosive
Orthophosphoric acid ≥85%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Catalog # BJ64957-1L
CAS Number:  
Corrosive
Orthophosphoric acid ≥85%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Catalog # BJ64957-1L
Supplier Number:  64957-1L
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. No.:
    BJ64957-1L
  • Supplier no.:
    64957-1L

Specifications

Specification Test Results

  • Sulfate (SO₄):
    Max. 5 ppm
  • Ammonium (NH₄):
    Max. 1 ppm
  • KMnO₄ reducing matter (as O):
    Max. 1 ppm
  • Assay:
    85.0 - 90.0 %
  • Density (D 20/4):
    1.691 - 1.706
  • Silver (Ag):
    Max. 1 ppb
  • Aluminium (Al):
    Max. 10 ppb
  • Arsenic (As):
    Max. 2 ppb
  • Gold (Au):
    Max. 1 ppb
  • Boron (B):
    Max. 10 ppb
  • Barium (Ba):
    Max. 1 ppb
  • Beryllium (Be):
    Max. 5 ppb
  • Bismuth (Bi):
    Max. 1 ppb
  • Calcium (Ca):
    Max. 50 ppb
  • Cadmium (Cd):
    Max. 1 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 1 ppb
  • Chromium (Cr):
    Max. 20 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 5 ppb
  • Dysprosium (Dy):
    Max. 0.5 ppb
  • Erbium (Er):
    Max. 0.5 ppb
  • Europium (Eu):
    Max. 0.5 ppb
  • Iron (Fe):
    Max. 100 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Gadolinium (Gd):
    Max. 1 ppb
  • Germanium (Ge):
    Max. 1 ppb
  • Hafnium (Hf):
    Max. 1 ppb
  • Mercury (Hg):
    Max. 0.5 ppb
  • Holmium (Ho):
    Max. 0.5 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lanthanum (La):
    Max. 0.5 ppb
  • Lithium (Li):
    Max. 1 ppb
  • Lutetium (Lu):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 5 ppb
  • Manganese (Mn):
    Max. 5 ppb
  • Molybdenum (Mo):
    Max. 5 ppb
  • Sodium (Na):
    Max. 50 ppb
  • Niobium (nb):
    Max. 5 ppb
  • Neodymium (nd):
    Max. 0.5 ppb
  • Nickel (Ni):
    Max. 20 ppb
  • Lead (Pb):
    Max. 1 ppb
  • Palladium (Pd):
    Max. 1 ppb
  • Praseodymium (Pr):
    Max. 0.5 ppb
  • Platinum (Pt):
    Max. 1 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhenium (Re):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 500 ppb
  • Selenium (Se):
    Max. 5 ppb
  • Samarium (Sm):
    Max. 0.5 ppb
  • Tin (Sn):
    Max. 2 ppb
  • Strontium (Sr):
    Max. 1 ppb
  • Tantalum (Ta):
    Max. 5 ppb
  • Terbium (Tb):
    Max. 0.5 ppb
  • Tellurium (Te):
    Max. 1 ppb
  • Thorium (Th):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 10 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Thulium (Tm):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 1 ppb
  • Tungsten (W):
    Max. 5 ppb
  • Yttrium (Y):
    Max. 0.5 ppb
  • Ytterbium (Yb):
    Max. 0.5 ppb
  • Zinc (Zn):
    Max. 10 ppb
  • Zirconium (Zr):
    Max. 5 ppb
  • Chloride (Cl):
    Max. 0.5 ppm
  • Nitrate (NO₃):
    Max. 0.5 ppm

Specification Test Results

About this item

 :
 :98 g/mol
Boiling Pt:158 °C (1.013 hPa)
Melting Pt:21 °C
Density:1.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340