ContactUsLinkComponent
Graphite Furnace AAS Optional Accessories, Agilent Technologies
Graphite Furnace AAS Optional Accessories, Agilent Technologies
Catalog # AG9910113100
undefined
Graphite Furnace AAS Optional Accessories, Agilent Technologies
Catalog # AG9910113100
Supplier Number:  9910113100
Specifications
  • Description:
    Carousel kit, high capacity, for GTA 120, for 130×1.1 ml vials and 5 bulk solutions
  • Cat. No.:
    AG9910113100
Specifications
About this item
Optional accessories for the Agilent graphite furnace AAS systems to help you improve productivity.
  • GTA viewing and fume extraction accessory mounts on sample compartment to remove fumes from the workhead of the GTA 120 and GTA 120 Zeeman, and improves viewing of the dispensing capillary for easier alignment
  • The high capacity carousel option for the PSD 120 furnace autosampler increases the sample capacity up to 130 samples, for improved productivity
  • The bubble free syringe update for the PSD-97/100 improves precision during extended furnace AA sequences, by eliminating bubble formation in the dispensing syringe
Agilent's optional accessories for graphite furnace AAS systems to help you improve productivity and ease of use. The GTA viewing and fume extraction accessory removes fumes and vapors that emerge from the graphite tube through the exhaust. It also improves viewing of the injection hole assists with alignment of the dispensing capillary of the PSD 120 furnace autosampler. It is compatible with the Agilent GTA 120 and GTA 120 Zeeman graphite furnace AAS. The high capacity carousel option increase sample capacity to 130 samples, with 5 central vessels for standards and modifier.;The bubble-free syringe update for the PSD-97/100 helps to eliminate bubble formation in the syringe during extended operation and improves long term precision.