Filters for MP-AES, Agilent Technologies
Catalog # AGG8001-64004
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Specifications
- Description:Inlet air filter kit, Agilent 4107 nitrogen generator
- Cat. No.:AGG8001-64004
Specifications
About this item
Cost-effective filters assure a supply of the highest purity gas/air to the MP-AES.
- Air inlet filter captures laboratory dust and debris to protect and prolong the instrument
- Replacement filter elements for the integrated air filter of the Agilent 4107 Nitrogen Generator remove large particles and residual oil vapor from the air supply to the nitrogen generator to prevent contamination
Agilent filters for the MP-AES replacement filter elements for the integrated air filter of the Agilent 4107 Nitrogen Generator and filter for the cooling air inlet on the MP-AES instruments.