Coolant fluids and Filters for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Polyclear PLUS coolant, strainer & air/water filters for the PolyScience chiller used with ICP-MS.
- Replacement air filter for the water chiller removes dust and large particles from the air supply to keep the system operating at optimum cooling capacity
- Replacement water strainer removes large particulates to prevent problems with clogging
- Chiller coolant levels should be inspected monthly to ensure the coolant level is adequate
- Annual maintenance is recommended - coolant should be completely drained and replaced, and the strainer and water filter should be cleaned
- Use genuine PolyClear PLUS coolant which includes algaecide and rust inhibitor for protection of your PolyScience chiller/recirculator and Agilent ICP-MS
Ensure that the 7700, 7800, 7900, 8800 and 8900 ICP-MS heat exchanger and G3292A chiller is operating with the optimum cooling capacity by using the recommended coolant, filters and supplies. Poly-clear Plus coolant is comprised of distilled water with algicide and rust inhibitor ensuring optimal chiller performance, while the strainer and fluid filter prevent particulates from clogging the cooling system on the instrument.