Contact Cylinders for PerkinElmer GFAAS Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Agilent contact cylinders equivalent to contact cylinders in PerkinElmer graphite furnace AAS system.
- Guaranteed to be compatible with PerkinElmer graphite furnace AAS systems
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Qualified on a genuine PerkinElmer system;Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
- Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
- Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent uses high-quality graphite and applies stringent quality controls to ensure contamination free and consistency of our contact cylinders for your PerkinElmer graphite furnace AA system. The standard THGA graphite contact cylinders, feature a modified injection port area that helps to minimize condensation and improve analytical performance with difficult matrices. The HGA contact cylinders have shaped graphite contacts to completely encase the graphite tube, ensuring the tube is positioned in a defined environment for uniform radiation conditions.