AdvanceBio Gly-X Technology, Agilent Technologies
Catalog # AGGX100
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Specifications
- Description:AdvanceBio Gly-X vacuum manifold spacer (formerly ProZyme)
- Cat. No.:AGGX100
Specifications
About this item
Gly-X sample preparation platform shortens time to analysis with fast, in-solution workflows.
The Agilent AdvanceBio Gly-X (formerly ProZyme) technology is a new generation of N-glycan sample preparation. Be ready for sample analysis faster with rapid (5-minute) enzymatic release of N-glycans and labeling with InstantDyes including the brightest in class InstantPC, or traditional reductive amination dyes. Innovative in-solution workflows combined with plate-based rapid cleanup enables sample prep in as little as 1 hour. Increase analytical sensitivity with a variety of dye options, while increasing sample throughput.