Vacuum and Chiller Fluids, Oils and Filters for PerkinElmer ICP-MS, Agilent Technologies
AG8003-0474
- Description:PerkinElmer ICP-MS Coolant fluid, for ELAN 9000/6×X00/DRC/NexION, 1 L
- Cat. No.:AG8003-0474
Agilent vacuum fluid (AVF) pump oils and chiller fluid for PerkinElmer NexION and ELAN ICP-MS.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Qualified on a genuine PerkinElmer system
- Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
- Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns, to help you get the most out of your PerkinElmer system
- In stock and available worldwide for 24- to 48-hour delivery (in most regions)
- Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
- Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent vacuum fluid (AVF) pump oils, filters and recirculator chiller fluids for PerkinElmer NexION 300/350 and ELAN 9000/6x00/DRCs ICP-MS instruments.
Achieve guaranteed performance with supplies for PE from Agilent that are equivalent to the PerkinElmer ICP-MS versions so you can use them with complete confidence.