ContactUsLinkComponent
Tubing and Other Supplies for PerkinElmer Autosamplers, Agilent Technologies
Tubing and Other Supplies for PerkinElmer Autosamplers, Agilent Technologies
Catalog # AG8003-0620
Tubing and Other Supplies for PerkinElmer Autosamplers, Agilent Technologies
Catalog # AG8003-0620
Supplier Number:  8003-0620
Specifications
  • Description:
    Capillary tubing assembly, PTFE, 1 m, 0.6 mm int.Ø, 2 fittings, red, for AS stainless steel probe, for FIAS, for AS-90/90A/90plus/91/93plus/S10 autosamplers
  • Cat. No.:
    AG8003-0620
Specifications
About this item
Capillary tubing and Other Supplies for PerkinElmer AS-90/90A/90plus/91/93plus/S10 autosamplers.
  • Guaranteed to be compatible with PerkinElmer AS-90/90A/90plus/91/93plus/S10 Autosamplers
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime;Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent capillary tubing is equivalent to PerkinElmer AA tubing, so you can be confident that it will integrate seamlessly with your PerkinElmer AS-90/90A/90plus/91/93plus/S10 autosampler.