Synthetic Chem Elut S Plates, Agilent Technologies
Catalog # AG5610-2003
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Specifications
- Volume:2 ml
- Description:Chem Elut S, 96-well plate, 200 µl sample
- Cat. No.:AG5610-2003
Specifications
About this item
Chem Elut S products offer superior SLE reproducibility with minimal method development.
- Controlled particle and pore size enable consistent flow and reproducibility
- Absence of shaking eliminates emulsion formation
- Effective removal of matrix, including salts and phospholipids
- Minimal extraction solvent required, leading to high analyte concentrations
- Reduced analyst-to-analyst variability compared to conventional LLE
Chem Elut S products enable reproducible SLE results with high data quality and sample capacity. Consistent pore and particle sizes allow for batch-to-batch repeatability and uniform flow. Emulsions are also avoided, because there is no need for shaking.
Compared to liquid-liquid extraction, Chem Elut S reduces analyst-to-analyst variability, improves matrix removal, and requires minimal extraction solvent, which leads to increased analyte concentration.