Face Shield, Full Length
Face shield, Clear, PPE, 9×13×2"
Catalog # 76549-684
Supplier: Mopec
Supplier #: BE197
Jump to:
Product Details & Documents
Reusable, easy to clean, face shields provide lightweight, full face protection and can be comfortably worn with eyeglasses. Constructed from latex free, fiberglass free and distortion free durable plastic.
Full-length reusable face shield offers lightweight, clear protection
- The window features an anti-fog and anti-static coating for all day wear
Foam headband relieves pressure and provides added comfort throughout.
Specifications
- Color:Clear
- Description:Face shield
- Dimensions:9×13×2"
- Material:PPE