ContactUsLinkComponent
No-Contact Infrared Thermometer, HT-820D, Mortech
No-Contact Infrared Thermometer, HT-820D, Mortech
Catalog # 76549-680
Supplier:  Mopec
undefined
No-Contact Infrared Thermometer, HT-820D, Mortech
Catalog # 76549-680
Supplier:  Mopec
Supplier Number:  BE195
Specifications
  • Description:
    No-contact infrared thermometer, HT-820D
  • Cat. No.:
    76549-680
Specifications
About this item
The HT-820D Body Infrared Thermometer is designed to be a screening device to measure body and forehead temperature without contact. The easily visible three-color alarm backlight display makes it easy to detect higher temperatures during temperature measurement.
  • Easily change units from °C to °F in 2 steps
  • Accuracy is ±0.3 °C (±0.6 °F)
  • Internal memory stores up to 34 readings for easy recall
  • Three color alarm backlight display visually show normal, slightly high, and serious temperatures