ContactUsLinkComponent
No Contact Infrared Thermometer, JRT200, Mortech
  76549-676
 :  Mopec
undefined
No Contact Infrared Thermometer, JRT200, Mortech
  76549-676
 :  Mopec
 :  BE193
  • Cat. No.:
    76549-676
  • Range:
    32...43 ºC
Featuring a large screen tri-color backlight, infrared contact measuring forehead temperature. Can store 32 sets of data.
  • 99.9 accurate temperature measurement
  • One second fast and convenient measurement
  • Non-contact ear or forehead measurement
  • Dual mode
  • Infrared sensor
  • Automatically shutdown during waiting
 : FDA and CE certified and approved.