Raised Dissection Table, Mortech
Catalog # 76549-614
Supplier: Mortech Manufacturing
Specifications
- Description:Raised dissection table
- Cat. No.:76549-614
Specifications
About this item
This raised dissection table is used to raise your dissection board above the body on your work surface.
- All polished stainless steel construction
- Heavy gauge material provides a very stable work surface
- Tray creased toward center drain
- Suitable for over the body placement
Drain has optional hose attachment to route flow to desired location.
Ordering information: Dissection Board sold separately.