Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Compact Scale, Digital, Mortech

Compact Scale, Digital, Mortech

16,000 g, 9.5×9.8×2.9"
Catalog # 76549-582
Supplier:   Mopec
Supplier #:  BB063
Compact Scale, Digital, Mortech
Catalog # 76549-582
Supplier:  Mopec
Compact Scale, Digital, Mortech
Compact Scale, Digital, Mortech

16,000 g, 9.5×9.8×2.9"

Price
$1,169.84
Log in for availability
Each
Product Details & Documents

Durable, fluid-resistant precision scale for use in all laboratory environments. Offers additional percent weighing mode and security features.

  • 1: 20 lb./10 kg calibration
  • LCD display
  • Rechargeable battery, 6 V (100 hour typical operation with 12-hour full charge), or 4 C batteries (not included)
Specifications
  • Dimensions:
    9.5×9.8×2.9"
  • Weighing capacity:
    16,000 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.