Order Entry
Puerto Rico
ContactUsLinkComponent
Scalpel Handle, Stainless Steel No. 4L, Premium Grade, Mortech
Scalpel Handle, Stainless Steel No. 4L, Premium Grade, Mortech
  76549-488
 :  Mopec
undefined
Scalpel Handle, Stainless Steel No. 4L, Premium Grade, Mortech
  76549-488
 :  Mopec
 :  AJ201

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Length:
    8.625"
  • Cat. No.:
    76549-488

 

 

High-quality #4 long scalpel handle, manufactured in Germany, can be used with #20 thru #23 blades. Stainless steel.

  • Premium grade stainless steel