Specifications
- Length:15.9 cm (6¼")
- Cat. No.:76549-124
Specifications
About this item
Ideal for delicate and light duty post-mortem dissection procedures.
- Serrated total perimeter grip
Available in a variety of thin to wide spaced curved prongs for gently isolating and retracting fascia, ligament tissue, nerve isolation, nerve teasing and spinal cord dissection. Serrated total perimeter grip assures secure tissue handling.