Cavity Mirror With Handle, Mortech
Catalog # 76549-016
Supplier: Mortech Manufacturing
Specifications
- Length:8"
- Cat. No.:76549-016
Specifications
About this item
Reverse cavity viewing for limited dissection areas such as oral, pediatric or small animal dissection specimens. Reduces finger probing for infectious cases.
- Stainless steel