Orders LinkContactUsLinkComponent
Forceps, CutMATE Starter Kit, Mortech
Forceps, CutMATE Starter Kit, Mortech
CutMATE forceps starter kit
Catalog # 76548-748
Supplier:   Mopec
Supplier #:  AB070
Forceps, CutMATE Starter Kit, Mortech
Catalog # 76548-748
Supplier:  Mopec
Forceps, CutMATE Starter Kit, Mortech
Forceps, CutMATE Starter Kit, Mortech
CutMATE forceps starter kit
Price
$2,658.74
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CutMATE forceps were developed to overcome the variability of tissue specimen thickness. They will allow cutting consistent tissue specimen thickness of 2, 3, or 4 mm.
  • 304 stainless steel
The unique 'trident-like' design of the 35 mm long ergonomically angled prongs, ensures uniform tissue thickness cutting of tissue lengths that will fit standard cassettes. The inside surface of the prongs is specially textured to provide the extra grip needed for stabilizing and slicing difficult and hard to hold specimens.
 : Includes (4) forceps, decontamination rack, and calibration device.
Specifications
  • Description:
    CutMATE forceps starter kit