Forceps, Semken Iris Tissue Dissecting, Mortech
5"
Catalog # 76548-688
Supplier: Mopec
: AB033
Jump to:
Product Details & Documents
Sharp interlocking teeth for secure grip with minimal tissue compression.
- Standard grade stainless steel
- Ribbed grip
- 1×2 teeth
Specifications
- Length:5"