Orders LinkContactUsLinkComponent
Dissecting Kit, Instructor, Mortech
  76548-516
 :   Mopec
 :  AA325
Dissecting Kit, Instructor, Mortech
  76548-516
 :  Mopec
Dissecting Kit, Instructor, Mortech
Dissecting Kit, Instructor, Mortech
$232.03
Each
Offers a variety of basic tools to accommodate most post-mortem dissection procedures.
  • Portable and versatile
  • 13 pieces plus case
 : Vinyl case kit contains one each of the following: dissection scissors, sharp/blunt, straight, 4.5" – AA007, dissection scissors, sharp/blunt, straight, 5.5" – AA008, general dissection forceps, serrated tips – AB003, dissection forceps, 1×2 teeth, 5.5" – AB011, dissection forceps, 1×2 Teeth, 6" – AB012, triple chain hook – AD029, section lifter, double ended 6" -AD040, scalpel, stainless steel -AJ130, scalpel, stainless steel, narrow – AJ131, teasing needle, metal handle, straight, teasing needle, metal handle, angled, eye dropper, ruler.