ContactUsLinkComponent
Scissors, Delicate, Double Curved, Sharp, Mortech
Scissors, Delicate, Double Curved, Sharp, Mortech
Catalog # 76548-496
Supplier:  Mopec
undefined
Scissors, Delicate, Double Curved, Sharp, Mortech
Catalog # 76548-496
Supplier:  Mopec
Supplier Number:  AA154
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    4.625"
  • Cat. No.:
    76548-496
Specifications
About this item
Finger-friendly ribbon handle. Extra sharp with single serrated edge. Double curvature of these scissors makes post-mortem dissection easier in hard to reach areas or where a clear view is not always available.
  • Premium grade stainless steel
  • Curved cutting edge
  • Sharp, one side of scissor edge is serrated