DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)
Catalog # 82022-774
Supplier: G-Biosciences
CAS Number:
Specifications
- Size:100 mg
- Cat. No.:82022-774
- Supplier no.:BC03
Specifications
Specification Test Results
- Spacer Arm (Å):19.9
- Reactive Toward:Sulfhydryls
- Membrane Permeable:not done
- Water Soluble:No
- Cleavable/ Reversible:Reducing Agents (Thiols)
Specification Test Results
About this item
:
:482.71 g/mol
CAS Number:141647-62-3
Formula:C₂₀H₂₆N₄O₂S₄
MDL Number:MFCD00153604