Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Behenic acid 85%, Technical Grade

Behenic acid 85%, Technical Grade

Behenic acid 85%, Technical Grade
Certificates
Behenic acid 85%, Technical Grade

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:13,01024
  • Boiling Pt:306 °C (60 mmHg)
  • Melting Pt:80…82 °C
  • Storage Temperature:Ambient
  • EINECS:204-010-8
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥80 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $167.00
    $167.00
    Each (250g)
       $167.00

    Supplier Number: 33481-2500
  • Glass bottle
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $424.00
    $424.00
    Each (1kg)
       $424.00

    Supplier Number: 33481-0010

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.