Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Adipic acid 99%

Corrosive
Adipic acid 99%
Certificates
Adipic acid 99%

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Hexanedioc acid, 1,6-Hexanedioic acid
  • CAS Number:124-04-9
  • Formula:HOOC(CH₂)₄COOH
  • MDL Number:MFCD00004420
  • Molecular Weight:146.14 g/mol
  • Merck Index:13,00163
  • Boiling Pt:337 °C (760 mmHg)
  • Melting Pt:151…153 °C
  • Flash Pt:196 °C
  • Storage Temperature:Ambient
  • EINECS:204-673-3
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 153 °C
  • Water:
    ≤0.2 %
  • Ash:
    ≤0.0002 %
  • Titration with NaOH:
    ≥99 %
  • Color of solution:
    ≤5 APHA (20g adipic acid + 20 g dist. H2O + 60 g 10% NH3 soln.)
  • Nitrate (NO3):
    ≤4 ppm
  • Iron (Fe):
    ≤0.4 ppm
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $34.00
    $34.00
    Each (500g)
       $34.00

    Supplier Number: 10281-5000
  • Plastic bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $43.40
    $43.40
    Each (1kg)
       $43.40

    Supplier Number: 10281-0010
  • Plastic bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g/mol)
    Each (2,500g/mol) - $74.10
    $74.10
    Each (2,500g/mol)
       $74.10

    Supplier Number: 10281-0025
  • Fibre drum
    25 kg
    Log in for availability
    Log in for availability
    Each (25kg)
    Each (25kg) - $429.00
    $429.00
    Each (25kg)
       $429.00

    Supplier Number: 10281-0250

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.