Virex® II 256 Empty Spray Bottle
Catalog # 76547-820
Supplier: DIVERSEY INC.
Specifications
- Capacity:946 ml (32 oz.)
- Description:Virex® II 256 Empty Spray Bottle
- Cat. No.:76547-820
Specifications
About this item
Empty spray bottle that can be filled with the disinfectant cleaner Virex® II 256.