Soft Care® Defend Antibacterial Soap
Catalog # 76547-792
Supplier: DIVERSEY INC.
Specifications
- Capacity:6x 1200 ml
- Description:Antimicrobal skin cleaner, refill
- Cat. No.:76547-792
Specifications
About this item
Soft Care® Defend(TM/MC) is a highly effective, mild, antibacterial soap.
- Safe, quick and effective formulation helps reduce cross-contamination
- Mild on skin, avoiding excessive cracking and dryness for even the most frequent users
- NSF E-2 certified for use in food handling establishments
- Formulated to ensure excellent removal of soil
- Skin compatibility (Patch) tested
This unique product combines the caring aspect of a gentle-on-skin cosmetic with antiseptic properties.
Formulated with chloroxylenol (PCMX), Soft Care® Defend(TM/MC) is effective at removing soils, rinses quickly and leaves no residue on skin.