Order Entry
Puerto Rico
ContactUsLinkComponent
Soft Care® Impact Foam Instant Hand Sanitizer 
Soft Care® Impact Foam Instant Hand Sanitizer 
Catalog # 76547-780
Supplier:  DIVERSEY INC.
undefined
Soft Care® Impact Foam Instant Hand Sanitizer 
Catalog # 76547-780
Supplier:  DIVERSEY INC.
Supplier Number:  100907873

Specifications

  • Capacity:
    6x 1200 ml
  • Description:
    Alcohol-based instant hand sanitizer, refill
  • Cat. No.:
    76547-780

Specifications

About this item

Soft Care® ImpactTM/MC is an alcohol based instant hand sanitizer enriched with skin conditioning emollients to keep skin soft and protect against irritation for even the most frequent users.

  • Contains 76% (v/v), 72% (w/w) ethyl alcohol, meeting APIC, PHAC, CDC and WHO Healthcare Personal Hand Wash guidelines
  • Skin softening emollients leave hands soft and with no sticky residue
  • Made with 96% biobased ingredients
  • Dye-free and fragrance-free formulas reduce potential risk of skin irritation and allergic reactions for users with sensitive skin
  • NSF E-3 certified for use in food handling establishments
  • Minimize the risk of sick leave resulting from cross contamination

Fragrance-free and dye-free.

Soft Care® ImpactTM/MC meets CDC, WHO, APIC and PHAC hand hygiene recommendations for Healthcare personnel hand rubs.