Soft Care® ImpactTM/MC Alcohol Liquid Instant Hand Sanitizer
76547-778
- Capacity:6x 1200 ml
- Description:Alcohol-based instant hand sanitizer, refill
- Cat. No.:76547-778
Soft Care® ImpactTM/MC is an alcohol based instant hand sanitizer enriched with skin conditioning emollients to keep skin soft and protect against irritation for even the most frequent users.
- Contains 76% (v/v), 72% (w/w) ethyl alcohol, meeting APIC, PHAC, CDC and WHO Healthcare Personal Hand Wash guidelines
- Skin softening emollients leave hands soft and with no sticky residue
- Made with 96% biobased ingredients
- Dye-free and fragrance-free formulas reduce potential risk of skin irritation and allergic reactions for users with sensitive skin
- NSF E-3 certified for use in food handling establishments
- Minimize the risk of sick leave resulting from cross contamination
Fragrance-free and dye-free.
Soft Care® ImpactTM/MC meets CDC, WHO, APIC and PHAC hand hygiene recommendations for Healthcare personnel hand rubs.