Order Entry
Puerto Rico
ContactUsLinkComponent
 
Antibac B, Highly-soluble Chlorinated Sanitizer
  76547-720
undefined
Antibac B, Highly-soluble Chlorinated Sanitizer
  76547-720
 :  08514210
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Size:
    1 x 350 lb
  • Cat. No.:
    76547-720
  • Packaging:
    Drum

 

 

Antibac B sanitizer is a rapid acting, highly soluble, granular, organic chlorinated solution.

  • Readily soluble for preparing stock solutions used in automatic sanitizing units.
  • Leaves no films on equiptment.
  • Can be used at 100 ppm for most sanitizing operations.

It is designed for use as a hard surface contact sanitizer in dairy plants, food processing plants, and pharmaceutical and cosmetics manufacturing facilities.

It can also be used for sanitizing commercial eggs.