Orders LinkContactUsLinkComponent
Oxivir® Tb Wipes
Oxivir® Tb Wipes
Supplier:   DIVERSEY INC.
Oxivir® Tb Wipes
Supplier:  DIVERSEY INC.
Certificates
Oxivir® Tb Wipes
Oxivir® Tb Wipes
2 Options Available Below
Product Details & Documents
Ready-to-use disinfectant cleaner wipes based on proprietary hydrogen peroxide (AHP®) technology to deliver fast, effective cleaning performance.
  • One-step cleaner disinfectant powered by AHP(R) technology
  • Effective against key pathogens - TB, MRSA, Norovirus and VRE
  • Kills HBV, HCV and HIV in 1 minute. Tuberculocidal in 5 minutes and Fungicidal in 3 minutes
  • No VOCs, no added fragrance and no NPEs. Environmentally responsible - active ingredient breaks down to oxygen and water
  • Compatible with most hard, non-porous surfaces
  • Convenient pre-wetted wipes make even the toughest job easier
Disinfects in 60 seconds.
Virucide, bactericide, tuberculocide, fungicide and non-food contact sanitizer.
Kills MRSA and Norovirus.
Meets bloodborne pathogen standards for decontaminating blood and body fluids.
Colorless with a characteristic scent.
Product Family Options
Product Information
  • No. of towels/wipes per container
    Packaging
    Availability
    Price
  • 4×160/pack
    Plastic dispensing can
    Log in for availability
    Log in for availability
    Carton of 4
    Carton of 4 - $339.20
    $339.20
    Carton of 4
       $339.20
    Supplier Number: 5627427
  • 4×160/pack
    Refill for Plastic can
    Log in for availability
    Log in for availability
    Carton of 4
    Carton of 4 - $275.67
    $275.67
    Carton of 4
       $275.67
    Supplier Number: 100823906