Virex® Tb Ready-to-Use Disinfectant Cleaner
76547-680
- Size:12 x 32 oz
- Cat. No.:76547-680
- Packaging:Plastic bottle
- Scent:Lemon
A ready-to-use, quaternary-based disinfectant that provides excellent cleaning and deodorizing in one step
- Kills HBV and HIV-1 (AIDS virus) in one minute. Provides broad spectrum kill of gram positive and gram negative microorganisms, including Tb, MSRA, VRE, MRSE, HBV, HIV-1, Canine parvovirus and many more
- Kills Tb in five minutes and all other bacteria in three minutes. Kills Norovirus in 30 seconds
- Ready-To-Use formula provides excellent, cost-effective cleaning and disinfection in one step
- Eliminates odors leaving surfaces smelling clean and fresh
Disinfects in three minutes. Bactericide, tuberculocide, virucide, fungicide. Kills Norovirus, VRE and MRSA.
Meets bloodborne pathogen standards for decontaminating blood and body fluids.
Colorless with a lemon scent.