


Tetrachlorophthalic anhydride 98%
Catalog # TS15779-2500
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Bottle
- Size:250 g
- Cat. No.:TS15779-2500
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:254 to 258 °C
- HPLC:≥97.5 %
- Color of alcohol solution:≤110 APHA (100 g in 70 ml methanol)
- Insoluble matter:≤0.5 % (10 g in 100 ml 1 M NaOH)
Specification Test Results
About this item
:
:285.9 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:254…258 °C
EINECS:204-171-4
UN:3077
:GHS08
:GHS05
:GHS09
CAS Number:117-08-8
Formula:C₈Cl₄O₃
MDL Number:MFCD00005920