

Allyl mercaptan 70% tech. remainder mainly sulfide
Catalog # TS15371-0250
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:25 g
- Cat. No.:TS15371-0250
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥65 %
Specification Test Results
About this item
:
:74.15 g/mol
Density:0.93 g/cm³
Flash Pt:18 °C
Storage Temperature:Refrigerator
EINECS:212-792-7
UN:1228
ADR:3,II
:GHS07
:GHS02
CAS Number:870-23-5
Formula:C₃H₆S
MDL Number:MFCD00004894