Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Maltol 99%

Maltol 99%

Irritant
Maltol 99%
Certificates
Maltol 99%

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
  • CAS Number:118-71-8
  • Formula:C₆H₆O₃
  • MDL Number:MFCD00006578
  • Molecular Weight:126.11 g/mol
  • Merck Index:13,05735
  • Boiling Pt:170 °C (100 mmHg)
  • Melting Pt:160…164 °C
  • Storage Temperature:Ambient
  • EINECS:204-271-8
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    160 to 164 °C
  • Water:
    ≤0.5 %
  • GC:
    ≥98.5 %
  • Arsenic (As):
    ≤3 ppm
  • Lead (Pb):
    ≤10 ppm
  • UV:
    at 220 nm A: ≤0.2
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤2 ppm
  • Acidity/alkalinity:
    ≤0.0001 meq/g
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $23.10
    $23.10
    Each (5g)
       $23.10

    Supplier Number: 12155-0050
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $45.80
    $45.80
    Each (25g)
       $45.80

    Supplier Number: 12155-0250
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $114.00
    $114.00
    Each (100g)
       $114.00

    Supplier Number: 12155-1000
  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $483.00
    $483.00
    Each (500g)
       $483.00

    Supplier Number: 12155-5000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.